Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCL3
Entrez Gene ID
823508
Gene Name
dicer-like 3
Gene Aliases
AT3G43920, ATDCL3, DICER-LIKE 3, dicer-like 3
Chromosome Location
Chr.3: 15753387 - 15760859 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161190.2 NP_001154662.2 22 - 23 4423 100
NM_001161191.3 NP_001154663.2 22 - 23 4541 100
NM_001339114.1 NP_001325812.1 22 - 23 4551 100
NM_001339115.1 NP_001325811.1 22 - 23 4554 100
NM_001339116.1 NP_001325813.1 17 - 18 3924 100
NM_114260.1 NP_189978.1 20 - 21 4276 100
GenBank mRNA DQ479972.1 - 20 - 21 4276 100

View Full Product Details