Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIT3
Entrez Gene ID
823557
Gene Name
nitrilase 3
Gene Aliases
AT3G44320, AtNIT3, NITRILASE 3, nitrilase 3
Chromosome Location
Chr.3: 15993259 - 15995811 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114300.4 NP_190018.1 2 - 3 476 82
GenBank mRNA BT002773.1 - 2 - 3 353 82
BT004345.1 - 2 - 3 316 82
U09959.1 - 2 - 3 337 82

View Full Product Details