Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEF
Entrez Gene ID
823559
Gene Name
hypothetical protein
Gene Aliases
AT3G44340, clone eighty-four
Chromosome Location
Chr.3: 16011547 - 16020336 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035731.2 NP_001030808.1 21 - 22 2945 128
NM_001339129.1 NP_001326943.1 20 - 21 3064 128
NM_001339130.1 NP_001326945.1 21 - 22 2965 128
NM_001339131.1 NP_001326944.1 21 - 22 2948 128
NM_114302.3 NP_566869.1 21 - 22 2935 128

View Full Product Details