Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAR8
Entrez Gene ID
823581
Gene Name
fatty acid reductase 8
Gene Aliases
AT3G44560, fatty acid reductase 8
Chromosome Location
Chr.3: 16151123 - 16154607 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339157.1 NP_001327123.1 8 - 9 1544 109
NM_114324.3 NP_190042.2 9 - 10 1379 109
GenBank mRNA DQ446732.1 - 9 - 10 1325 109
EU280153.1 - 9 - 10 1325 109

View Full Product Details