Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP71
Entrez Gene ID
823585
Gene Name
cyclophilin71
Gene Aliases
AT3G44600, AtCYP71, cyclophilin 71, cyclophilin71
Chromosome Location
Chr.3: 16164935 - 16169304 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114328.4 NP_190046.2 11 - 12 1767 75
GenBank mRNA AY062653.1 - 11 - 12 1725 75
BT008382.1 - 11 - 12 1664 75

View Full Product Details