Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL9
Entrez Gene ID
823623
Gene Name
ribosomal protein L9
Gene Aliases
AT3G44890, ribosomal protein L9
Chromosome Location
Chr.3: 16386395 - 16388723 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114358.4 NP_190075.1 6 - 7 737 131
GenBank mRNA Z11129.1 - - 636 131

View Full Product Details