Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX11
Entrez Gene ID
823626
Gene Name
cation/H+ exchanger 11
Gene Aliases
AT3G44920, ATCHX11, cation/H+ exchanger 11
Chromosome Location
Chr.3: 16396925 - 16399924 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114361.3 NP_190078.2 2 - 3 1441 129

View Full Product Details