Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl48
Entrez Gene ID
823636
Gene Name
serine carboxypeptidase-like 48
Gene Aliases
AT3G45010, serine carboxypeptidase-like 48
Chromosome Location
Chr.3: 16466144 - 16468989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114370.4 NP_190087.1 7 - 8 1465 75
GenBank mRNA AY050427.1 - 7 - 8 1319 75
AY093993.1 - 7 - 8 1281 75

View Full Product Details