Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAG2
Entrez Gene ID
828471
Gene Name
vacuolar ATP synthase subunit G2
Gene Aliases
AT4G23710, F9D16.180, F9D16_180, VACUOLAR ATP SYNTHASE SUBUNIT G2, VATG2, VHA-G2, vacuolar ATP synthase subunit G2
Chromosome Location
Chr.4: 12349926 - 12351624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118502.3 NP_194102.1 3 - 4 445 89
GenBank mRNA AK227689.1 - 3 - 4 282 89
BT003107.1 - 2 - 3 176 89

View Full Product Details