Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBR5
Entrez Gene ID
814997
Gene Name
indole-3-butyric acid response 5
Gene Aliases
AT2G04550, DSPTP1E, DUAL SPECIFICITY PROTEIN PHOSPHATASE 1E, T1O3.4, T1O3_4, indole-3-butyric acid response 5
Chromosome Location
Chr.2: 1588193 - 1590166 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126486.6 NP_178534.2 2 - 3 692 126
NM_201689.3 NP_973418.2 2 - 3 525 126
GenBank mRNA AY337455.1 - 2 - 3 478 126
BT004283.1 - 2 - 3 481 126
BT005609.1 - 2 - 3 347 126

View Full Product Details