Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZAP1
Entrez Gene ID
815035
Gene Name
zinc-dependent activator protein-1
Gene Aliases
AT2G04880, ATWRKY1, F1O13.1, F1O13_1, WRKY1, zinc-dependent activator protein-1
Chromosome Location
Chr.2: 1717833 - 1720526 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126520.4 NP_178565.1 2 - 3 1077 131
NM_179605.3 NP_849936.1 3 - 4 1060 131
GenBank mRNA AB493546.1 - 2 - 3 748 131
AF442389.1 - 1 - 2 820 131
AF442390.1 - 2 - 3 748 131
BT029167.1 - 2 - 3 748 131
X92976.1 - - 941 131

View Full Product Details