Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS11
Entrez Gene ID
815065
Gene Name
vacuolar protein sorting 11
Gene Aliases
AT2G05170, ATVPS11, F5G3.7, F5G3_7, vacuolar protein sorting 11
Chromosome Location
Chr.2: 1869853 - 1873997 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126544.3 NP_027676.1 2 - 3 551 91
GenBank mRNA AF436824.1 - 2 - 3 519 91
BT002698.1 - 2 - 3 384 91

View Full Product Details