Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOD1
Entrez Gene ID
815152
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT2G05990, ENOYL-ACP REDUCTASE, ENOYL-ACP REDUCTASE 1, ENR1, MOSAIC DEATH 1, T6P5.19, T6P5_19
Chromosome Location
Chr.2: 2322215 - 2325197 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126612.3 NP_565331.1 8 - 9 1013 124
NM_179609.1 NP_849940.1 8 - 9 1003 124
GenBank mRNA AY056192.1 - 8 - 9 974 124
AY113962.1 - 7 - 8 816 124

View Full Product Details