The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084415.2 | NP_001077884.1 | 3 - 4 | 307 | 86 |
NM_126619.4 | NP_178662.1 | 3 - 4 | 480 | 86 | |
NM_201702.2 | NP_973431.1 | 2 - 3 | 428 | 86 | |
GenBank mRNA | AF370582.1 | - | 2 - 3 | 219 | 86 |
AF410322.1 | - | 3 - 4 | 264 | 86 | |
AK317250.1 | - | 2 - 3 | 429 | 86 | |
AY097367.1 | - | 2 - 3 | 219 | 86 |