Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ELF8
- Entrez Gene ID
-
815177
- Gene Name
- binding protein
- Gene Aliases
-
AT2G06210, EARLY FLOWERING 8, F5K7.3, F5K7_3, VERNALIZATION INDEPENDENCE 6, VIP6
- Chromosome Location
-
Chr.2: 2428903 - 2436687 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_126631.7 |
NP_178674.7 |
8 - 9 |
1049 |
90 |
View Full Product Details