Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAS1
Entrez Gene ID
815275
Gene Name
cycloartenol synthase 1
Gene Aliases
AT2G07050, CYCLOARTENOL SYNTHASE 1, T4E14.16, T4E14_16, cycloartenol synthase 1
Chromosome Location
Chr.2: 2924360 - 2930751 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335311.1 NP_001325220.1 2 - 3 850 111
NM_126681.3 NP_178722.1 2 - 3 652 111
GenBank mRNA AY094394.1 - 2 - 3 565 111
BT001118.1 - 2 - 3 383 111

View Full Product Details