Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA6
Entrez Gene ID
815329
Gene Name
H[+]-ATPase 6
Gene Aliases
AHA6, AT2G07560, F9A16.7, F9A16_7, H(+)-ATPase 6
Chromosome Location
Chr.2: 3169908 - 3174102 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126721.3 NP_178762.1 3 - 4 1197 116

View Full Product Details