Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRCK3
Entrez Gene ID
815625
Gene Name
calmodulin-binding receptor-like cytoplasmic kinase 3
Gene Aliases
AT2G11520, F14P14.15, F14P14_15, calmodulin-binding receptor-like cytoplasmic kinase 3
Chromosome Location
Chr.2: 4618545 - 4621722 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126855.4 NP_565351.1 3 - 4 836 140
GenBank mRNA AF367339.1 - 3 - 4 721 140
AY093990.1 - 2 - 3 493 140

View Full Product Details