Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MGDC
Entrez Gene ID
815657
Gene Name
monogalactosyldiacylglycerol synthase type C
Gene Aliases
AT2G11810, ATMGD3, F7E22.4, F7E22_4, MGD3, MONOGALACTOSYL DIACYLGLYCEROL SYNTHASE 3, MONOGALACTOSYL DIACYLGLYCEROL SYNTHASE 3, MONOGALACTOSYLDIACYLGLYCEROL SYNTHASE, monogalactosyldiacylglycerol synthase type C
Chromosome Location
Chr.2: 4743162 - 4746813 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124829.2 NP_001118301.1 2 - 3 554 82
NM_001335374.1 NP_001323965.1 2 - 3 711 82
NM_001335375.1 NP_001323966.1 1 - 2 223 82
NM_126865.5 NP_565352.1 2 - 3 711 82
GenBank mRNA AB047398.1 - 2 - 3 507 82
BT026357.1 - 2 - 3 485 82
KJ138917.1 - 2 - 3 485 82

View Full Product Details