Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGT
Entrez Gene ID
815822
Gene Name
alanine:glyoxylate aminotransferase
Gene Aliases
AGT1, ALANINE:GLYOXYLATE AMINOTRANSFERASE, ALANINE:GLYOXYLATE AMINOTRANSFERASE 1, AT2G13360, F14O4.7, F14O4_7, L-serine:glyoxylate aminotransferase, SGAT, alanine:glyoxylate aminotransferase
Chromosome Location
Chr.2: 5538740 - 5541479 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335388.1 NP_001318216.1 3 - 4 1065 79
NM_126925.5 NP_178969.1 3 - 4 1135 79
NM_179620.2 NP_849951.1 3 - 4 1006 79
GenBank mRNA AB048945.1 - 3 - 4 916 79
AF063901.1 - 3 - 4 927 79
AY096498.1 - 3 - 4 925 79
AY114010.1 - 2 - 3 857 79

View Full Product Details