Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GONST1
Entrez Gene ID
815850
Gene Name
golgi nucleotide sugar transporter 1
Gene Aliases
AT2G13650, T10F5.19, T10F5_19, golgi nucleotide sugar transporter 1
Chromosome Location
Chr.2: 5687316 - 5690795 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202595.1 NP_001189524.1 6 - 7 867 107
NM_001335395.1 NP_001318219.1 5 - 6 1000 107
NM_001335396.1 NP_001324974.1 4 - 5 1318 107
NM_001335397.1 NP_001324975.1 3 - 4 660 107
NM_001335398.1 NP_001324973.1 5 - 6 926 107
NM_126944.5 NP_565357.1 5 - 6 991 107
NM_179621.3 NP_849952.1 5 - 6 991 107
GenBank mRNA BT030001.1 - 4 - 5 736 107
KR265320.1 - 4 - 5 736 107

View Full Product Details