Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH6B2
Entrez Gene ID
815903
Gene Name
aldehyde dehydrogenase 6B2
Gene Aliases
AT2G14170, T22C12.10, T22C12_10, aldehyde dehydrogenase 6B2, aldehyde dehydrogenase 6B2
Chromosome Location
Chr.2: 5977272 - 5981899 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084419.1 NP_001077888.1 1 - 2 237 114
NM_126989.2 NP_179032.1 1 - 2 237 114

View Full Product Details