Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD28
Entrez Gene ID
838000
Gene Name
IQ-domain 28
Gene Aliases
AT1G14380, F14L17.15, F14L17_15, IQ-domain 28
Chromosome Location
Chr.1: 4918010 - 4921682 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035969.2 NP_001031046.1 3 - 4 441 104
NM_001332114.1 NP_001319001.1 3 - 4 786 104
NM_001332115.1 NP_001323290.1 3 - 4 605 104
NM_001332116.1 NP_001323291.1 3 - 4 666 104
NM_001332117.1 NP_001323289.1 3 - 4 535 104
NM_101305.4 NP_563950.1 3 - 4 579 104
NM_202095.3 NP_973824.1 3 - 4 493 104
GenBank mRNA AK117145.1 - 3 - 4 493 104
BT005935.1 - 2 - 3 321 104

View Full Product Details