Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD28
Entrez Gene ID
838000
Gene Name
IQ-domain 28
Gene Aliases
AT1G14380, F14L17.15, F14L17_15, IQ-domain 28
Chromosome Location
Chr.1: 4918010 - 4921682 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035969.2 NP_001031046.1 6 - 7 2083 77
NM_001332114.1 NP_001319001.1 7 - 8 2242 77
NM_101305.4 NP_563950.1 6 - 7 2221 77
GenBank mRNA AK117145.1 - 6 - 7 2135 77

View Full Product Details