Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP10
Entrez Gene ID
839112
Gene Name
tubby like protein 10
Gene Aliases
AT1G25280, AtTLP10, F4F7.33, F4F7_33, tubby like protein 10
Chromosome Location
Chr.1: 8863768 - 8866894 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123881.1 NP_001117353.1 3 - 4 1180 117
NM_102338.3 NP_173899.1 4 - 5 1171 117
NM_202180.1 NP_973909.1 3 - 4 1099 117
GenBank mRNA AF487271.1 - 3 - 4 585 117
BT001997.1 - 3 - 4 1099 117

View Full Product Details