Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THA2
Entrez Gene ID
819608
Gene Name
threonine aldolase 2
Gene Aliases
AT3G04520, threonine aldolase 2
Chromosome Location
Chr.3: 1217174 - 1219847 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202878.1 NP_001189807.1 2 - 3 560 111
NM_001337534.1 NP_001327633.1 1 - 2 570 111
NM_111323.4 NP_566228.1 2 - 3 636 111
GenBank mRNA AF325033.2 - 2 - 3 524 111
AK175345.1 - 2 - 3 496 111
AY085456.1 - 2 - 3 496 111
AY091093.1 - 2 - 3 548 111
AY123033.1 - 1 - 2 458 111

View Full Product Details