Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THA2
Entrez Gene ID
819608
Gene Name
threonine aldolase 2
Gene Aliases
AT3G04520, threonine aldolase 2
Chromosome Location
Chr.3: 1217174 - 1219847 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202878.1 NP_001189807.1 7 - 8 980 89
NM_001337534.1 NP_001327633.1 6 - 7 993 89
NM_111323.4 NP_566228.1 7 - 8 1059 89
GenBank mRNA AF325033.2 - 7 - 8 947 89
AK175345.1 - 7 - 8 919 89
AY085456.1 - 7 - 8 919 89
AY091093.1 - 7 - 8 971 89
AY123033.1 - 6 - 7 881 89

View Full Product Details