Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPCK2
- Entrez Gene ID
-
819609
- Gene Name
- phosphoenolpyruvate carboxylase kinase 2
- Gene Aliases
-
AT3G04530, ATPPCK2, PEPCK2, PHOSPHOENOLPYRUVATE CARBOXYLASE KINASE 2, T27C4.19, T27C4_19, phosphoenolpyruvate carboxylase kinase 2
- Chromosome Location
-
Chr.3: 1221422 - 1222744 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111324.4 |
NP_566229.1 |
1 - 2 |
918 |
86 |
View Full Product Details