Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA16
Entrez Gene ID
819633
Gene Name
indoleacetic acid-induced protein 16
Gene Aliases
AT3G04730, F7O18.22, F7O18_22, indoleacetic acid-induced protein 16
Chromosome Location
Chr.3: 1288155 - 1290891 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111345.3 NP_187124.1 1 - 2 680 127
GenBank mRNA AF332400.1 - 1 - 2 204 127
AF375420.1 - 1 - 2 387 127
AK229447.1 - 1 - 2 387 127
AY087195.1 - 1 - 2 391 127
BT000857.1 - 1 - 2 204 127
U49072.1 - 1 - 2 370 127

View Full Product Details