Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLDP2
Entrez Gene ID
819730
Gene Name
phospholipase D P2
Gene Aliases
AT3G05630, F18C1.10, F18C1_10, PDLZ2, PHOSPHOLIPASE D, PHOSPHOLIPASE D ZETA 2, PLDZETA2, phospholipase D P2
Chromosome Location
Chr.3: 1635194 - 1640357 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111436.6 NP_187214.2 3 - 4 577 96
GenBank mRNA AM182458.1 - 3 - 4 503 96
AY305003.1 - 3 - 4 450 96

View Full Product Details