Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMPA-7
Entrez Gene ID
819741
Gene Name
importin alpha
Gene Aliases
AT3G05720, F18C1.1, F18C1_1, importin alpha isoform 7
Chromosome Location
Chr.3: 1687583 - 1691821 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337604.1 NP_001319482.1 4 - 5 580 85
NM_111446.2 NP_187223.1 4 - 5 515 85
GenBank mRNA DQ446636.1 - 4 - 5 495 85

View Full Product Details