Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RECQI1
Entrez Gene ID
819743
Gene Name
RECQ helicase l1
Gene Aliases
AT3G05740, F18C1.21, RECQ helicase l1
Chromosome Location
Chr.3: 1698079 - 1701467 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111448.3 NP_187225.2 1 - 2 211 66
GenBank mRNA AJ404470.1 - 1 - 2 178 66

View Full Product Details