Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RECQI1
Entrez Gene ID
819743
Gene Name
RECQ helicase l1
Gene Aliases
AT3G05740, F18C1.21, RECQ helicase l1
Chromosome Location
Chr.3: 1698079 - 1701467 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337605.1 NP_001327100.1 7 - 8 1475 73
NM_111448.3 NP_187225.2 8 - 9 1595 73
GenBank mRNA AJ404470.1 - 8 - 9 1562 73

View Full Product Details