Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCHR12
Entrez Gene ID
819772
Gene Name
SNF2/Brahma-type chromatin-remodeling protein CHR12
Gene Aliases
AT3G06010, CHR12, F2O10.3, F2O10_3, chromatin remodeling 12
Chromosome Location
Chr.3: 1802141 - 1807530 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111475.3 NP_187252.2 1 - 2 462 63

View Full Product Details