Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- P4H2
- Entrez Gene ID
-
819804
- Gene Name
- P4H isoform 2
- Gene Aliases
-
AT-P4H-2, AT3G06300, F24P17.24, F24P17_24, P4H isoform 2, PROLYL 4-HYDROXYLASE, prolyl 4-hydroxylase 2
- Chromosome Location
-
Chr.3: 1907582 - 1910063 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111505.4 |
NP_566279.1 |
6 - 7 |
764 |
95 |
View Full Product Details