The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_111524.4 | NP_187300.3 | 1 - 2 | 533 | 82 |
GenBank mRNA | AF039406.1 | - | 1 - 2 | 379 | 82 |
AY035017.1 | - | 1 - 2 | 393 | 82 | |
AY059083.1 | - | 1 - 2 | 250 | 82 |