Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBR3
Entrez Gene ID
819865
Gene Name
acyl-CoA dehydrogenase-like protein
Gene Aliases
AT3G06800, AT3G06810, IBA-RESPONSE 3
Chromosome Location
Chr.3: 2146243 - 2150851 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111561.5 NP_187337.2 11 - 12 1878 92
GenBank mRNA AY091014.1 - 11 - 12 1641 92
BT002365.1 - 11 - 12 1587 92

View Full Product Details