Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX13
Entrez Gene ID
819945
Gene Name
peroxin 13
Gene Aliases
ABERRANT PEROXISOME MORPHOLOGY 2, APM2, AT3G07560, peroxin 13
Chromosome Location
Chr.3: 2411321 - 2413587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111634.6 NP_187412.1 4 - 5 968 100
GenBank mRNA AY075689.1 - 4 - 5 765 100
AY093745.1 - 4 - 5 691 100

View Full Product Details