Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCN1
Entrez Gene ID
819979
Gene Name
Immunoglobulin E-set superfamily protein
Gene Aliases
AT3G07880, SUPERCENTIPEDE1
Chromosome Location
Chr.3: 2513925 - 2515983 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111667.4 NP_187445.1 3 - 4 774 94
GenBank mRNA AF412276.1 - 3 - 4 532 94
AK118247.1 - 3 - 4 699 94
AY085508.1 - 3 - 4 728 94
BT005578.1 - 3 - 4 524 94

View Full Product Details