Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPKKK6
Entrez Gene ID
819989
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
AT3G07980, MAP3K EPSILON PROTEIN KINASE 2, MAP3KE2, mitogen-activated protein kinase kinase kinase 6
Chromosome Location
Chr.3: 2543603 - 2551360 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337756.1 NP_001326459.1 11 - 12 1206 109
NM_001337757.1 NP_001326460.1 7 - 8 762 109
NM_111677.4 NP_187455.1 11 - 12 1242 109

View Full Product Details