Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL27
Entrez Gene ID
819990
Gene Name
serine carboxypeptidase-like 27
Gene Aliases
AT3G07990, serine carboxypeptidase-like 27
Chromosome Location
Chr.3: 2552394 - 2554921 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111678.3 NP_187456.1 1 - 2 523 128
GenBank mRNA BT015083.1 - 1 - 2 432 128
BT015904.1 - 1 - 2 373 128

View Full Product Details