Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRE1
Entrez Gene ID
828504
Gene Name
trehalase 1
Gene Aliases
AT4G24040, ATTRE1, T19F6.30, T19F6_30, trehalase 1
Chromosome Location
Chr.4: 12488040 - 12491298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118536.4 NP_194135.1 3 - 4 1149 77
GenBank mRNA AK118407.1 - 3 - 4 1006 77
BT010732.1 - 3 - 4 947 77

View Full Product Details