Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF8
Entrez Gene ID
828515
Gene Name
growth-regulating factor 8
Gene Aliases
AT4G24150, AtGRF8, AtMIN10, T19F6.140, T19F6_140, growth-regulating factor 8
Chromosome Location
Chr.4: 12535972 - 12539576 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341644.1 NP_001329478.1 5 - 6 943 68
NM_001341645.1 NP_001329477.1 3 - 4 1210 68
NM_118547.2 NP_194146.1 5 - 6 1018 68

View Full Product Details