Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO13
Entrez Gene ID
828526
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT4G24250, ATMLO13, MILDEW RESISTANCE LOCUS O 13, T22A6.80, T22A6_80
Chromosome Location
Chr.4: 12574719 - 12577583 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341655.1 NP_001328763.1 3 - 4 457 68
NM_118558.4 NP_567697.1 3 - 4 457 68
GenBank mRNA AF369574.1 - 3 - 4 421 68
AK221618.1 - 3 - 4 410 68
BT026373.1 - 3 - 4 391 68

View Full Product Details