Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIPK8
Entrez Gene ID
828542
Gene Name
CBL-interacting protein kinase 8
Gene Aliases
AT4G24400, ATCIPK8, CBL-interacting protein kinase 8, PKS11, PROTEIN KINASE 11, SNF1-RELATED PROTEIN KINASE 3.13, SnRK3.13, T22A6.230, T22A6_230
Chromosome Location
Chr.4: 12616979 - 12620714 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084973.1 NP_001078442.1 4 - 5 526 134
NM_118573.4 NP_194171.1 4 - 5 803 134
GenBank mRNA AF290193.1 - 4 - 5 403 134
BT000958.1 - 4 - 5 403 134

View Full Product Details