Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBP16
- Entrez Gene ID
-
828558
- Gene Name
- ubiquitin-specific protease 16
- Gene Aliases
-
AT4G24560, F22K18.240, F22K18_240, ubiquitin-specific protease 16
- Chromosome Location
-
Chr.4: 12679200 - 12684976 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118589.3 |
NP_567705.1 |
7 - 8 |
2625 |
108 |
View Full Product Details