Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22D
Entrez Gene ID
828598
Gene Name
HVA22 homologue D
Gene Aliases
ARABIDOPSIS THALIANA HVA22 HOMOLOGUE D, AT4G24960, ATHVA22D, F13M23.100, F13M23_100, HVA22 homologue D
Chromosome Location
Chr.4: 12827856 - 12829303 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203901.1 NP_001190830.1 1 - 2 175 72
NM_118628.4 NP_567713.1 1 - 2 255 72
GenBank mRNA AF141662.1 - 1 - 2 174 72
AY059750.1 - 1 - 2 174 72
AY086850.1 - 1 - 2 174 72
AY113978.1 - 1 - 2 51 72

View Full Product Details