Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIN2
Entrez Gene ID
828626
Gene Name
RPM1 interacting protein 2
Gene Aliases
AT4G25230, F24A6.70, F24A6_70, RPM1 interacting protein 2
Chromosome Location
Chr.4: 12923682 - 12929274 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341724.1 NP_001328533.1 2 - 3 499 132
NM_118655.4 NP_194253.2 2 - 3 613 132
NM_179221.2 NP_849552.1 1 - 2 827 132
GenBank mRNA AY072178.1 - 1 - 2 614 132

View Full Product Details