Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XTH14
Entrez Gene ID
828687
Gene Name
xyloglucan endotransglucosylase/hydrolase 14
Gene Aliases
AT4G25820, ATXTH14, F14M19.100, F14M19_100, XTR9, xyloglucan endotransglucosylase/hydrolase 14, xyloglucan endotransglycosylase 9
Chromosome Location
Chr.4: 13130291 - 13131860 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118714.4 NP_194312.1 2 - 3 532 117
GenBank mRNA AF093672.1 - 2 - 3 515 117
AY093183.1 - 2 - 3 526 117
BT003385.1 - 2 - 3 474 117

View Full Product Details