Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUM6
Entrez Gene ID
828694
Gene Name
pumilio 6
Gene Aliases
APUM6, AT4G25880, F14M19.160, F14M19_160, pumilio 6
Chromosome Location
Chr.4: 13154708 - 13159382 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036643.2 NP_001031720.1 8 - 9 2436 71
NM_001341767.1 NP_001329827.1 7 - 8 2394 71
NM_001341768.1 NP_001329828.1 7 - 8 2394 71
NM_118721.3 NP_567733.1 8 - 9 2715 71
NM_202889.3 NP_974618.2 8 - 9 2516 71
GenBank mRNA AF360298.1 - 8 - 9 2531 71
AK319148.1 - 7 - 8 2885 71
AY133866.1 - 7 - 8 2394 71

View Full Product Details